Learn More
Hampton Research CRYSTAL SCREEN NO18 185ML
SDP
Catalog No. 501427
Encompass
HR2-910-18/Crystal Screen #18 (B6), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 20% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-142-7 Supplier Hampton Research Supplier No. HR291018
HR2-910-18/Crystal Screen #18 (B6), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 20% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.