Learn More
Hampton Research CRYSTAL SCREEN NO19 185ML
SDP
Catalog No. 501428
Encompass
HR2-910-19/Crystal Screen #19 (B7), 185 ml - 0.2 M Ammonium acetate, 0.1 M TRIS hydrochloride pH 8.5, 30% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-142-8 Supplier Hampton Research Supplier No. HR291019
HR2-910-19/Crystal Screen #19 (B7), 185 ml - 0.2 M Ammonium acetate, 0.1 M TRIS hydrochloride pH 8.5, 30% v/v 2-Propanol. Please note items from this supplier are non-returnable.