Learn More
Hampton Research CRYSTAL SCREEN NO20 185ML
SDP
Catalog No. 501429
Encompass
HR2-910-20/Crystal Screen #20 (B8), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 25% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-142-9 Supplier Hampton Research Supplier No. HR291020
HR2-910-20/Crystal Screen #20 (B8), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 25% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.