Learn More
Hampton Research CRYSTAL SCREEN NO22 185ML
SDP
Catalog No. 501431
Encompass
HR2-910-22/Crystal Screen #22 (B10), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-143-1 Supplier Hampton Research Supplier No. HR291022
HR2-910-22/Crystal Screen #22 (B10), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.