Learn More
Hampton Research CRYSTAL SCREEN NO26 185ML
SDP
Catalog No. 501435
Encompass
HR2-910-26/Crystal Screen #26 (C2), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-143-5 Supplier Hampton Research Supplier No. HR291026
HR2-910-26/Crystal Screen #26 (C2), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.