Learn More
Hampton Research CRYSTAL SCREEN NO28 185ML
SDP
Supplier:  Hampton Research HR291028
Encompass
HR2-910-28/Crystal Screen #28 (C4), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 30% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-143-7