Learn More
Hampton Research CRYSTAL SCREEN NO29 185ML
SDP
Supplier:  Hampton Research HR291029
Encompass
HR2-910-29/Crystal Screen #29 (C5), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.8 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-143-8