Learn More
Hampton Research CRYSTAL SCREEN NO30 185ML
SDP
Catalog No. 501439
Encompass
HR2-910-30/Crystal Screen #30 (C6), 185 ml - 0.2 M Ammonium sulfate, 30% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-143-9 Supplier Hampton Research Supplier No. HR291030
HR2-910-30/Crystal Screen #30 (C6), 185 ml - 0.2 M Ammonium sulfate, 30% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.