Learn More
Hampton Research CRYSTAL SCREEN NO31 185ML
SDP
Catalog No. 501440
Encompass
HR2-910-31/Crystal Screen #31 (C7), 185 ml - 0.2 M Ammonium sulfate, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-144-0 Supplier Hampton Research Supplier No. HR291031
HR2-910-31/Crystal Screen #31 (C7), 185 ml - 0.2 M Ammonium sulfate, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.