Learn More
Hampton Research CRYSTAL SCREEN NO34 185ML
SDP
Catalog No. 501443
Encompass
HR2-910-34/Crystal Screen #34 (C10), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 2.0 M Sodium formate. Please note items from this supplier are non-returnable.
Catalog No. 50-144-3 Supplier Hampton Research Supplier No. HR291034
HR2-910-34/Crystal Screen #34 (C10), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 2.0 M Sodium formate. Please note items from this supplier are non-returnable.