Learn More
Hampton Research CRYSTAL SCREEN NO35 185ML
SDP
Catalog No. 501444
Encompass
HR2-910-35/Crystal Screen #35 (C11), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.8 M Sodium phosphate monobasic monohydrate, 0.8 M Potassium phosphate monobasic. Please note items from this supplier are non-returnable.
Catalog No. 50-144-4 Supplier Hampton Research Supplier No. HR291035
HR2-910-35/Crystal Screen #35 (C11), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.8 M Sodium phosphate monobasic monohydrate, 0.8 M Potassium phosphate monobasic. Please note items from this supplier are non-returnable.