Learn More
Hampton Research CRYSTAL SCREEN NO36 185ML
SDP
Catalog No. 501445
Encompass
HR2-910-36/Crystal Screen #36 (C12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 8% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-144-5 Supplier Hampton Research Supplier No. HR291036
Add to Cart
Add to Cart
HR2-910-36/Crystal Screen #36 (C12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 8% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.