Learn More
Hampton Research CRYSTAL SCREEN NO37 185ML
SDP
Catalog No. 501446
Encompass
HR2-910-37/Crystal Screen #37 (D1), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 8% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-144-6 Supplier Hampton Research Supplier No. HR291037
HR2-910-37/Crystal Screen #37 (D1), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 8% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.