Learn More
Hampton Research CRYSTAL SCREEN NO38 185ML
SDP
Catalog No. 501447
Encompass
HR2-910-38/Crystal Screen #38 (D2), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.4 M Sodium citrate tribasic dihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-144-7 Supplier Hampton Research Supplier No. HR291038
HR2-910-38/Crystal Screen #38 (D2), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.4 M Sodium citrate tribasic dihydrate. Please note items from this supplier are non-returnable.