Learn More
Hampton Research CRYSTAL SCREEN NO39 185ML
SDP
Catalog No. 501448
Encompass
HR2-910-39/Crystal Screen #39 (D3), 185 ml - 0.1 M HEPES sodium pH 7.5, 2% v/v Polyethylene glycol 400, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-144-8 Supplier Hampton Research Supplier No. HR291039
HR2-910-39/Crystal Screen #39 (D3), 185 ml - 0.1 M HEPES sodium pH 7.5, 2% v/v Polyethylene glycol 400, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.