Learn More
Hampton Research CRYSTAL SCREEN NO40 185ML
SDP
Catalog No. 501449
Encompass
HR2-910-40/Crystal Screen #40 (D4), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 20% v/v 2-Propanol, 20% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-144-9 Supplier Hampton Research Supplier No. HR291040
HR2-910-40/Crystal Screen #40 (D4), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 20% v/v 2-Propanol, 20% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.