Learn More
Hampton Research CRYSTAL SCREEN NO42 185ML
SDP
Catalog No. 501451
Encompass
HR2-910-42/Crystal Screen #42 (D6), 185 ml - 0.05 M Potassium phosphate monobasic, 20% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-145-1 Supplier Hampton Research Supplier No. HR291042
HR2-910-42/Crystal Screen #42 (D6), 185 ml - 0.05 M Potassium phosphate monobasic, 20% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.