Learn More
Hampton Research CRYSTAL SCREEN NO45 185ML
SDP
Catalog No. 501454
Encompass
HR2-910-45/Crystal Screen #45 (D9), 185 ml - 0.2 M Zinc acetate dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 18% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-145-4 Supplier Hampton Research Supplier No. HR291045
HR2-910-45/Crystal Screen #45 (D9), 185 ml - 0.2 M Zinc acetate dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 18% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.