Learn More
Hampton Research CRYSTAL SCREEN NO46 185ML
SDP
Catalog No. 501455
Encompass
HR2-910-46/Crystal Screen #46 (D10), 185 ml - 0.2 M Calcium acetate hydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 18% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-145-5 Supplier Hampton Research Supplier No. HR291046
HR2-910-46/Crystal Screen #46 (D10), 185 ml - 0.2 M Calcium acetate hydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 18% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.