Learn More
Hampton Research CRYSTAL SCREEN NO47 185ML
SDP
Catalog No. 501456
Encompass
HR2-910-47/Crystal Screen #47 (D11), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-145-6 Supplier Hampton Research Supplier No. HR291047
HR2-910-47/Crystal Screen #47 (D11), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.