Learn More
Hampton Research CRYSTAL SCREEN NO48 185ML
SDP
Catalog No. 501457
Encompass
HR2-910-48/Crystal Screen #48 (D12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 2.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.
Catalog No. 50-145-7 Supplier Hampton Research Supplier No. HR291048
HR2-910-48/Crystal Screen #48 (D12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 2.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.