Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, 0.1% AQ, Vion Biosciences
SCI_ED

Catalog No. S02514 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02514 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING:This product can expose you to Crystal violet, which is known to the State of California to cause cancer.
Catalog No. S02514 Supplier Aldon Corporation Supplier No. VNCC0456500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING:This product can expose you to Crystal violet, which is known to the State of California to cause cancer.

Visualize proteins and DNA in living cells with histology grade crystal violet 0.1% aqueous stain.

  • Visualize proteins and DNA in living cells with histology grade crystal violet 0.1% aqueous stain.

Chemical Identifiers

CAS 532-32-1 , 7732-18-5 , 548-62-9
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00011750
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym Crystal Violet, Water Solution
PubChem CID Data not available.
ChEBI CHEBI:41688
IUPAC Name sodium benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point ~0°C (32°F in water)
Boiling Point ~100°C (212°F in water)
CAS Min % 0.2%, 99.7%, 0.1%
Concentration 0.10%
Dye Content 0.10%
Extinction Coefficient 87000 cm-1 M-1 (at 590 nm)
Identification Crystal Violet, 0.1% Aqueous Solution
CAS Max % 99.7%, 0.2%, 0.1%
Infrared Spectrum Yes
Lambda Max 590 nm
Molar Absorption Coefficient 87000 M-1cm-1
Odor No odor.
Packaging HDPE Bottle
Specific Gravity ~1.0 g/mL (water)
Assay Percent Range 0.10%
Vapor Pressure 14 mm Hg in water)
Certificate of Analysis Yes
UN Number Not regulated.
Decomposition Information Data not available.
Density ~1.0 g/mL (water)
pH Color Changes Data not available.
Solvent or Matrix Water
Color Dark Violet/Blue
Solubility Information Complete in water.
pH Data not available.
Quantity 500 mL
Physical Form Data not available.
Percent Purity 0.10%
Grade Data not available.
Concentration or Composition (by Analyte or Components) 0.1% Aqueous
Chemical Name or Material Crystal Violet
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Acute Toxicity, Oral
Hazard Statement Eye Irritation
ShelfLife 3 years
DOTInformation Not regulated.
EINECSNumber 208-953-6
RTECSNumber BO9000000
Recommended Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.