Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, 0.2% w/v in Acetic Acid, Reagents

Catalog No. 01335558
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-558 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-558 Supplier Reagents Holdings Llc Supplier No. CC263500500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystal Violet, 0.2% w/v in Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7 , 548-62-9
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Gentian Violet, Basic Violet 3
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Boiling Point 118°C
Packaging Glass Bottle
Specific Gravity 1.05
Color Dark Blue
pH <2
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.2%
Grade Reagent
Chemical Name or Material Crystal Violet, 0.2% w/v in Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.