Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, 1% Aqueous, Innovating Science™
SCI_ED

Catalog No. S031074 Shop All Aldon Products
Change view
Click to view available options
Volume (Metric):
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S031074 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! :This product can expose you to Crystal violet, which is known to the State of California to cause cancer.
Catalog No. S031074 Supplier Aldon Corporation Supplier No. IS12146
Add to Cart
Add to Cart
WARNING! :This product can expose you to Crystal violet, which is known to the State of California to cause cancer.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.