Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, 1% w/v, Reagents

Catalog No. 01336552
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-552 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-552 Supplier Reagents Holdings Llc Supplier No. C7113500100C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystal Violet, 1% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 548-62-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Gentian Violet, Basic Violet 3
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.0068
Color Violet
Quantity 100 mL
Physical Form Liquid
Percent Purity 1%
Grade Reagent
Chemical Name or Material Crystal Violet, 1% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.