Learn More
Sigma Aldrich Fine Chemicals Biosciences Crystal Violet, 548-62-9, MFCD00011750, 50 g
Catalog No. NC0827019
Encompass
Linear Formula: C25H30N3Cl, Molecular Weight: 407.98, ACS reagent, ≥90.0% anhydrous basis, Synonym: Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B. Crystal violet has been used to determine cell viability. It has been used in the staining of cells for the quantification of cellular migration.
Catalog No. NC0827019 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C615850G
Linear Formula: C25H30N3Cl, Molecular Weight: 407.98, ACS reagent, ≥90.0% anhydrous basis, Synonym: Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B. Crystal violet has been used to determine cell viability. It has been used in the staining of cells for the quantification of cellular migration.