Learn More
Medchemexpress LLC Crystal Violet | 548-62-9 | MFCD00011750 | 96.0-100.5% | 10 G
SDP
Catalog No. 5000406795
Encompass_Preferred
Crystal Violet is a triphenyl-methane, alkaline dye that binds to DNA in the nucleus of a cell, staining it a deep purple. It is commonly used for Gram staining to classify bacteria, or for cell or histological staining.
  • Triphenyl-methane alkaline dye
  • Binds to DNA in the nucleus of a cell
  • Stains cells a deep purple
  • Used for Gram staining to classify bacteria
  • Suitable for cell or histological staining
Catalog No. 50-004-06795 Supplier Medchemexpress LLC Supplier No. HYB0324A10G
Add to Cart
Add to Cart
Crystal Violet is a triphenyl-methane, alkaline dye that binds to DNA in the nucleus of a cell, staining it a deep purple. It is commonly used for Gram staining to classify bacteria, or for cell or histological staining.
  • Triphenyl-methane alkaline dye
  • Binds to DNA in the nucleus of a cell
  • Stains cells a deep purple
  • Used for Gram staining to classify bacteria
  • Suitable for cell or histological staining