Learn More
Bioworld Crystal Violet, 100 g
Catalog No. 50255831
Encompass_Preferred
A component of Gram staining in microorganisms and cell lines. Product is supplied in glass bottles.Specifications:Certified GradeSynonym: Basic Violet 3; Methyl Violet 10BEM (590nm, H2O) >93,500Moisture <7.5%Ash <1.5%Lead <0.003%Zinc <0.05%HS Code : 32041300RTECS Number: BO9000000
Catalog No. 50-255-831 Supplier Bioworld Supplier No. 403400161
A component of Gram staining in microorganisms and cell lines. Product is supplied in glass bottles.Specifications:Certified GradeSynonym: Basic Violet 3; Methyl Violet 10BEM (590nm, H2O) >93,500Moisture <7.5%Ash <1.5%Lead <0.003%Zinc <0.05%HS Code : 32041300RTECS Number: BO9000000