Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal violet, ACS, ≥90%, MP Biomedicals™

Catalog No. ICN15251190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15251190 500 g
ICN15251150 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15251190 Supplier MP Biomedicals, Inc Supplier No. 0215251190

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Hexamethylpararosaniline chloride, Methyl Violet 10B
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Assay Percent Range ≥90%
Formula Weight 408
Color Green
Quantity 500 g
Physical Form Powder
Chemical Name or Material Crystal violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.