Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, ACS Reagent (Hexamethylpararosaniline Chloride), Reagents

Catalog No. 01336768
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-768 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-768 Supplier Reagents Holdings Llc Supplier No. C610870010C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystal Violet, ACS Reagent (Hexamethylpararosaniline Chloride), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.99
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Gentian Violet, Basic Violet 3
IUPAC Name tris[4-(dimethylamino)phenyl]methylium chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Boiling Point 561°C
Packaging Glass Bottle
Specific Gravity 0.981
Color Dark Green
Quantity 10 g
Physical Form Solid
Grade ACS
Chemical Name or Material Crystal Violet, ACS Reagent (Hexamethylpararosaniline Chloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.