Learn More
Description
Crystal violet has been used to determine cell viability. It has been used in the staining of cells for the quantification of cellular migration.
Specifications
Specifications
| CAS | 548-62-9 |
| Molecular Weight (g/mol) | 407.98 g/mol |
| Grade | ACS |
| Color | Black to Green |
| Quantity | 50 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.