Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, ACS, Sigma-Aldrich

Catalog No. 01259054 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-054 100 g
01-258-816 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-054 Supplier MilliporeSigma Supelco Supplier No. C6158100G
Add to Cart
Add to Cart

Crystal violet has been used to determine cell viability. It has been used in the staining of cells for the quantification of cellular migration.

TRUSTED_SUSTAINABILITY

Specifications

CAS 548-62-9
Molecular Weight (g/mol) 407.98 g/mol
Grade ACS
Color Black to Green
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.