Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Crystal Violet, Certified, C.I. DcC-38 25 GR
SDP

Catalog No. 50980339
Encompass_Preferred

(Basic Violet 3; Gentian Violet; Methyl Violet 10B; Hexamethyl Violet)
CAS #548-62-9 C₂₅H₃₀N₃Cl F.W. 407.99
Solubility: 9.0% Water; 8.75% Alcohol; 7.5% Cellosolve; 7.0% Glycol; 0.0% Xylene

For Gram positive/Gram negative bacteria, filaments, Holzer's method for glial fibers, Amyloid in human tissue and chromatin and nucleoli in plant tissue.

Catalog No. 50-980-339 Supplier Electron Microscopy Sciences Supplier No. 12785
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Basic Violet 3; Gentian Violet; Methyl Violet 10B; Hexamethyl Violet)
CAS #548-62-9 C₂₅H₃₀N₃Cl F.W. 407.99
Solubility: 9.0% Water; 8.75% Alcohol; 7.5% Cellosolve; 7.0% Glycol; 0.0% Xylene

For Gram positive/Gram negative bacteria, filaments, Holzer's method for glial fibers, Amyloid in human tissue and chromatin and nucleoli in plant tissue.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.