Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Crystal Violet, Certified Biological Stain
SCI_ED

Catalog No. S25274B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Solid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25274B 25 g Solid
S25274 1 g Solid
S25274A 10 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25274B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Gentian Violet, Gram Stain
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Linear Formula C25H30ClN3
Formula Weight 408
Packaging Solid
Density 1g/mL
Color Green
Quantity 25 g
Chemical Name or Material Crystal Violet, Certified Biological Stain
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.