Learn More
Description
Crystal violet is used as an active component, primary stain, of Gram stain for differentiation of Gram-negative versus Gram-positive bacteria.
It has been used to check cell viability. It has also been used for the staining of cells to study cell migration and invasion.
Specifications
Specifications
| CAS | 548-62-9 |
| Color | Black to Green |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 407.98 g/mol |
| Grade | Certified Biological Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.