Learn More
Sigma Aldrich Fine Chemicals Biosciences Crystal Violet for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0) | 548-62-9 | MFCD00011750 | 100G
Catalog No. 501806555
Encompass
Crystal Violet for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0) | Mol Wt: 407.98 | 548-62-9 | MFCD00011750 | 100G
Catalog No. 50-180-6555 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 61135100G
Crystal Violet for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0) | Mol Wt: 407.98 | 548-62-9 | MFCD00011750 | 100G