Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet Indicator, 0.1% (w/v) in Glacial Acetic Acid, for Nonaqueous Titrations, Ricca Chemical
SDP

Catalog No. 24794 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2479-4 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2479-4 Supplier Ricca Chemical Company Supplier No. 24794
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Nonaqueous Titrations

Chemical Identifiers

CAS 64-19-7 , 548-62-9
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 98.0, 0.09
CAS Max % 100.0, 0.1
Concentration 0.1% (w/v)
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
Color Blue to Purple/Violet
pH <2
Quantity 120 mL
Physical Form Liquid
Percent Purity 0.1% W/V
Grade Indicator
Chemical Name or Material Crystal Violet Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.