Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Crystal Violet, pure, high purity biological stain

Catalog No. AC229641000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC229641000 100 g
AC229645000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229641000 Supplier Thermo Scientific Chemicals Supplier No. 229641000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Crystal Violet
Melting Point 215.0°C
CAS 548-62-9, 90-94-8
Infrared Spectrum Authentic
Loss on Drying 9% max. (110°C)
Packaging Glass bottle
Molecular Formula C25H30ClN3
MDL Number MFCD00011750
Beilstein 13, 756
Merck Index 15, 4430
Color Green
Synonym Basic Violet 3, C.I. 42555, Gentian Violet
Solubility Information Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
Quantity 100 g
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Physical Form Powder or Crystals
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.986
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.99
Percent Purity ≥96.0% (TiCl3-titration) (on dried substance)
Grade Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.