Learn More
Crescent Chemical Co Inc CRYSTAL VIOLET RESEARCH GRADE
SDP
Catalog No. 50247787
Encompass_Preferred
Crystal Violet research grade This dye is chemically homogeneous and well-defined, particularly recommended for Flemming and gram-staining because of reproducible results. Commercial preparations usually contain components of reddish tinge like methyl violet 2B.
Catalog No. 50-247-787 Supplier Crescent Chemical Co Inc Supplier No. SE27335.01
Crystal Violet research grade This dye is chemically homogeneous and well-defined, particularly recommended for Flemming and gram-staining because of reproducible results. Commercial preparations usually contain components of reddish tinge like methyl violet 2B.