Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet Solution R, EP Reagent 1022901, Ricca Chemical
SDP

Catalog No. R2484000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R2484000500 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2484000500 Supplier Ricca Chemical Company Supplier No. R2484000500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-19-7 , 548-62-9
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 98.0, 0.46
CAS Max % 100.0, 0.48
Packaging Amber Glass Bottle
Color Blue to Purple/Violet
pH <1
Quantity 500 mL
Physical Form Liquid
Grade Indicator
Chemical Name or Material Crystal Violet Solution, Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.