Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet Solution, Sigma-Aldrich

Catalog No. 01258844 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-844 500 mL
01-259-056 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-844 Supplier MilliporeSigma Supelco Supplier No. V5265500ML
Add to Cart
Add to Cart

Crystal violet is the primary stain in Gram staining. It infiltrates the peptidoglycan-rich cell wall of Gram-positive bacteria, and forms entrapped complexes with Gram's iodine that retain the purple color post-decolorization. Thus, Gram-positive bacteria can be differentiated from Gram-negative bacteria whose peptidoglycan layer is considerably thinner.

TRUSTED_SUSTAINABILITY

Specifications

CAS 548-62-9
Molecular Weight (g/mol) 407.98 g/mol
Color Dark Purple
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.