Learn More
Volu Sol Crystal Violet Stain, 16 oz / 500 mL, Volu-Sol™
SDP
Catalog No. NC9745557 Shop All Volu Sol Products
Encompass_Preferred
Crystal Violet, component of the Gram Stain Kit.  This solution acts as the primary stain in Gram procedures
Catalog No. NC9745557 Supplier Volu Sol Supplier No. VGS016
Crystal Violet, component of the Gram Stain Kit.  This solution acts as the primary stain in Gram procedures