Learn More
Mitegen Goniometer base type B5-R, MicroRT compatible; package of 1000
SDP
Catalog No. 501939519
Encompass_Preferred
Reusable Glue-Free Magnetic Bases/Caps for holding crystal loops. (SPINE Style) Fully compatible with ALS style robotic sample changers. Unique design compatible with MicroRT™ capillaries for room temperature screening and crystal dehydration studies. Can be used with all crystal mounts/loops, including MicroMounts™ , MicroLoops™ , MicroGrippers™ and MicroMesh™ . *Reusable Bases are compatible only with MiTeGen Brand Mounts and Loops.
Catalog No. 50-193-9519 Supplier Mitegen Supplier No. GBB5R1000
Add to Cart
Add to Cart
Request a Quote
Reusable Glue-Free Magnetic Bases/Caps for holding crystal loops. (SPINE Style) Fully compatible with ALS style robotic sample changers. Unique design compatible with MicroRT™ capillaries for room temperature screening and crystal dehydration studies. Can be used with all crystal mounts/loops, including MicroMounts™ , MicroLoops™ , MicroGrippers™ and MicroMesh™ . *Reusable Bases are compatible only with MiTeGen Brand Mounts and Loops.