Learn More
Mitegen Goniometer base type B1, MicroRT compatible; package of 1000
SDP
Catalog No. 501939463
Encompass_Preferred
Magnetic Bases/Caps for holding crystal loops. Fully compatible with SSRL/SAM style robotic sample changers. Unique design compatible with MicroRT™ capillaries for room temperature screening and crystal dehydration studies. Can be used with all crystal mounts/loops, including MicroMounts™ , MicroLoops™ , MicroGrippers™ and MicroMesh™ . Style available in both MiTeGen’s patented Reusable Glue-free design or in the regular Glue-required Design
Catalog No. 50-193-9463 Supplier Mitegen Supplier No. GBB11000
Add to Cart
Add to Cart
Request a Quote
Magnetic Bases/Caps for holding crystal loops. Fully compatible with SSRL/SAM style robotic sample changers. Unique design compatible with MicroRT™ capillaries for room temperature screening and crystal dehydration studies. Can be used with all crystal mounts/loops, including MicroMounts™ , MicroLoops™ , MicroGrippers™ and MicroMesh™ . Style available in both MiTeGen’s patented Reusable Glue-free design or in the regular Glue-required Design