Learn More
Electron Microscopy Sciences 509, Color: White. Softening: 125°F. Flow point: 130°F. Viscosity: 500 cps., 1/EA
SDP
Catalog No. 502090586
Encompass_Preferred
A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. Color: White. Softening: 125°F . Flow point: 130°F . Viscosity: 500 cps.
Catalog No. 50-209-0586 Supplier Electron Microscopy Sciences Supplier No. 5040101HMP
A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. Color: White. Softening: 125°F . Flow point: 130°F . Viscosity: 500 cps.