Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Crystalbond™ 590 Adhesive Stick, Brown 225 GR
SDP

Catalog No. 50997402
Encompass_Preferred

A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. For precise high purity work, offers maximum adhesion to subminiature parts.
Color: Brown
Softening: 257°F (125°C)
Flow point: 302°F (150°C)
Viscosity: 9,000 cps

Catalog No. 50-997-402 Supplier Electron Microscopy Sciences Supplier No. 5040202
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. For precise high purity work, offers maximum adhesion to subminiature parts.
Color: Brown
Softening: 257°F (125°C)
Flow point: 302°F (150°C)
Viscosity: 9,000 cps

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.