Learn More
Electron Microscopy Sciences Crystal Clear™ 200 Water Liquid Plastics 1 GL
SDP
Catalog No. 5031100
Encompass_Preferred
The Crystal Clear Series of polyurethane resins were designed for applications that require absolute clarity. These are low viscosity resins that cure at room temperature with negligible shrinkage to a hard plastic. Cured products are UV resistant and are not brittle. Applications include potting, encapsulating, making prototype models, and lenses, object duplicating etc.
CAUTION: This product is NOT for home use, for research and industrial use only.
Each kit consists of 1 gallon resin A and adequate curing agent B 10 minute pot life. Casting thickness .5 to 3 in.
Pot Life: 10 min
Full Cure: 16 hrs
Casting thickness: ½-3" (1.27-7.62cm)
Catalog No. 50-311-00 Supplier Electron Microscopy Sciences Supplier No. 24200
The Crystal Clear Series of polyurethane resins were designed for applications that require absolute clarity. These are low viscosity resins that cure at room temperature with negligible shrinkage to a hard plastic. Cured products are UV resistant and are not brittle. Applications include potting, encapsulating, making prototype models, and lenses, object duplicating etc.
CAUTION: This product is NOT for home use, for research and industrial use only.
Each kit consists of 1 gallon resin A and adequate curing agent B 10 minute pot life. Casting thickness .5 to 3 in.
Pot Life: 10 min
Full Cure: 16 hrs
Casting thickness: ½-3" (1.27-7.62cm)