Learn More
Electron Microscopy Sciences Crystal Clear™ 204 Water Liquid Plastics 1 GL
SDP
Supplier:  Electron Microscopy Sciences 24204
Encompass_Preferred
The Crystal Clear Series of polyurethane resins were designed for applications that require absolute clarity. These are low viscosity resins that cure at room temperature with negligible shrinkage to a hard plastic. Cured products are UV resistant and are not brittle. Applications include potting, encapsulating, making prototype models, and lenses, object duplicating etc.
CAUTION: This product is NOT for home use, for research and industrial use only.
Each kit consists of 1 gallon resin A and adequate curing agent B 48 hour pot life. Casting thickness 3 to 6 in.
Pot Life: 120 min
Full Cure: 48 hrs
Casting thickness: 3-6" (7.62-15.24cm)
Catalog No. 50-311-01