Learn More
Molecular Dimensions CRYSTALCLENE 18MM SQUARE (100)
SDP
Catalog No. 501038564
Encompass
CrystalClene Slips 18mm Square (100) Plastic coverslides. Do not require silanization. Unbreakable, chemical & solvent resistant. Supplied protected in a sandwich film or interleaved. Guaranteed dust-free. Guaranteed RNase-free.
Catalog No. 50-103-8564 Supplier Molecular Dimensions Supplier No. MD416
CrystalClene Slips 18mm Square (100) Plastic coverslides. Do not require silanization. Unbreakable, chemical & solvent resistant. Supplied protected in a sandwich film or interleaved. Guaranteed dust-free. Guaranteed RNase-free.