Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CrystalDrop™ Lids for CrystalQuick™ Microplates

Catalog No. p-7075380 Shop All Greiner Bio-One Products
Click to view available options
Format:
Pre-greased
Standard

Catalog No. 07-000-558

Supplier: Greiner Bio-One 609050

For use with CrystalQuick™ microplates. Greiner Bio-One CrystalDrop™ lids enables individual sealing of 96-wells.

  • 192-wells helps to place sample drops at predefined positions
  • Printed grid facilitates automated monitoring of crystallisation drops
  • Helps to conduct 288 sitting drop and 192 hanging drop experiments in parallel (total: 480 crystal
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Dimensions (L x W x H) 127.35 x 84.8 x 6.0 mm
Sterility Non-sterile
No. of Wells 96
No. per Case 40
Format Pre-greased
Plate Compatibility CrystalQuick Microplates

For research use only. Not for use in diagnostic procedures, unless otherwise indicated

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.